Merck
  • iRegNet3D: three-dimensional integrated regulatory network for the genomic analysis of coding and non-coding disease mutations.
iRegNet3D: three-dimensional integrated regulatory network for the genomic analysis of coding and non-coding disease mutations.
Genome biology (2017-01-20)
Siqi Liang, Nathaniel D Tippens, Yaoda Zhou, Matthew Mort, Peter D Stenson, David N Cooper, Haiyuan Yu
ZUSAMMENFASSUNG
The mechanistic details of most disease-causing mutations remain poorly explored within the context of regulatory networks. We present a high-resolution three-dimensional integrated regulatory network (iRegNet3D) in the form of a web tool, where we resolve the interfaces of all known transcription factor (TF)-TF, TF-DNA and chromatin-chromatin interactions for the analysis of both coding and non-coding disease-associated mutations to obtain mechanistic insights into their functional impact. Using iRegNet3D, we find that disease-associated mutations may perturb the regulatory network through diverse mechanisms including chromatin looping. iRegNet3D promises to be an indispensable tool in large-scale sequencing and disease association studies.