Merck
Dimethyl Fumarate Controls the NRF2/DJ-1 Axis in Cancer Cells: Therapeutic Applications.
Molecular cancer therapeutics (2017-01-11)
Nathaniel Edward Bennett Saidu, Gaëlle Noé, Olivier Cerles, Luc Cabel, Niloufar Kavian-Tessler, Sandrine Chouzenoux, Mathilde Bahuaud, Christiane Chéreau, Carole Nicco, Karen Leroy, Bruno Borghese, François Goldwasser, Frédéric Batteux, Jérôme Alexandre
ZUSAMMENFASSUNG
The transcription factor NRF2 (NFE2L2), regulates important antioxidant and cytoprotective genes. It enhances cancer cell proliferation and promotes chemoresistance in several cancers. Dimethyl fumarate (DMF) is known to promote NRF2 activity in noncancer models. We combined
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human PARK7