Merck
Imbalance of endogenous prostanoids in moderate-to-severe asthma.
Allergology international : official journal of the Japanese Society of Allergology (2016-07-19)
Masaya Takemura, Akio Niimi, Hisako Matsumoto, Tetsuya Ueda, Masafumi Yamaguchi, Hirofumi Matsuoka, Makiko Jinnai, Kian Fan Chung, Michiaki Mishima
ZUSAMMENFASSUNG
Inhalation studies suggested "protective" roles of exogenous prostaglandin E Mild (n = 41) or moderate-to-severe (19) asthmatics and 27 normal controls were examined for pulmonary function (FEV Each prostanoid level did not differ among the three groups. Sputum number of bronchial epithelial cells was greater in moderate-to-severe asthmatics than in the other two groups, suggesting epithelial desquamation. Levels of prostaglandin F An imbalance in production, breakdown, or both between prostaglandin E