Merck
  • Soluble CD14 inhibits contractile function and insulin action in primary adult rat cardiomyocytes.
Soluble CD14 inhibits contractile function and insulin action in primary adult rat cardiomyocytes.
Biochimica et biophysica acta (2016-11-07)
Sabrina Overhagen, Marcel Blumensatt, Pia Fahlbusch, Daniella Herzfeld de Wiza, Heidi Müller, Bujar Maxhera, Payam Akhyari, D Margriet Ouwens
ZUSAMMENFASSUNG
Epicardial adipose tissue (EAT) from patients with type 2 diabetes (T2D) is characterized by monocyte infiltrations and displays an elevated release of the monocyte marker soluble cluster of differentiation 14 (sCD14) versus EAT from patients without T2D. We propose that an increased abundance of sCD14 in EAT from patients with T2D may impair the function and insulin sensitivity of the adjacent cardiomyocytes. To examine this, primary adult rat cardiomyocytes were incubated with increasing concentrations of sCD14 in the presence and absence of the co-receptor lipopolysaccharide (LPS), and analyzed for effects on determinants of contractile function, activation of inflammation signalling and insulin action. Exposing cardiomyocytes to sCD14 increased the phosphorylation of the stress kinases p38 and extracellular-signal regulated kinase (ERK). In contrast, insulin-mediated phosphorylation of Akt on Thr308 and Ser473 was inhibited. Furthermore, sCD14 impaired sarcomere shortening and cytosolic Ca
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Glyceraldehyd, ≥90% (GC)