Merck
  • mTORC1-independent TFEB activation via Akt inhibition promotes cellular clearance in neurodegenerative storage diseases.
mTORC1-independent TFEB activation via Akt inhibition promotes cellular clearance in neurodegenerative storage diseases.
Nature communications (2017-02-07)
Michela Palmieri, Rituraj Pal, Hemanth R Nelvagal, Parisa Lotfi, Gary R Stinnett, Michelle L Seymour, Arindam Chaudhury, Lakshya Bajaj, Vitaliy V Bondar, Laura Bremner, Usama Saleem, Dennis Y Tse, Deepthi Sanagasetti, Samuel M Wu, Joel R Neilson, Fred A Pereira, Robia G Pautler, George G Rodney, Jonathan D Cooper, Marco Sardiello
ZUSAMMENFASSUNG
Neurodegenerative diseases characterized by aberrant accumulation of undigested cellular components represent unmet medical conditions for which the identification of actionable targets is urgently needed. Here we identify a pharmacologically actionable pathway that controls cellular clearance via Akt modulation of transcription factor EB (TFEB), a master regulator of lysosomal pathways. We show that Akt phosphorylates TFEB at Ser467 and represses TFEB nuclear translocation independently of mechanistic target of rapamycin complex 1 (mTORC1), a known TFEB inhibitor. The autophagy enhancer trehalose activates TFEB by diminishing Akt activity. Administration of trehalose to a mouse model of Batten disease, a prototypical neurodegenerative disease presenting with intralysosomal storage, enhances clearance of proteolipid aggregates, reduces neuropathology and prolongs survival of diseased mice. Pharmacological inhibition of Akt promotes cellular clearance in cells from patients with a variety of lysosomal diseases, thus suggesting broad applicability of this approach. These findings open new perspectives for the clinical translation of TFEB-mediated enhancement of cellular clearance in neurodegenerative storage diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
ANTI-FLAG® in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution