Merck
  • Unraveling the cytotoxicity and metabolic pathways of binary natural deep eutectic solvent systems.
Unraveling the cytotoxicity and metabolic pathways of binary natural deep eutectic solvent systems.
Scientific reports (2017-02-02)
Yves Paul Mbous, Maan Hayyan, Won Fen Wong, Chung Yeng Looi, Mohd Ali Hashim
ZUSAMMENFASSUNG
In this study, the anticancer potential and cytotoxicity of natural deep eutectic solvents (NADESs) were assessed using HelaS3, PC3, A375, AGS, MCF-7, and WRL-68 hepatic cell lines. NADESs were prepared from choline chloride, fructose, or glucose and compared with an N,N-diethyl ethanolammonium chloride:triethylene glycol DES. The NADESs (98 ≤ EC
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triethylenglycol, ReagentPlus®, 99%