- Effect of hydraulic retention time on suppression of methanogens during a continuous biohydrogen production process using molasses wastewater.
Effect of hydraulic retention time on suppression of methanogens during a continuous biohydrogen production process using molasses wastewater.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2016-09-10)
Jeong Hee Yun, Kyung-Suk Cho
PMID27610651
ZUSAMMENFASSUNG
This study was undertaken to investigate the reduction of the hydraulic retention time (HRT) to decrease methane generation and recover hydrogen production during the long-term operation of biohydrogen production in a continuous stirred tank reactor (CSTR) using molasses wastewater. Reduction of HRT can be a simple and economic method to immediately control unfavorable methane generated during continuous operation of a hydrogen production system. The steady-state performance of the CSTR showed a hydrogen content of 41.3 ± 3.30% and a hydrogen production rate (HPR) of 63.7 ± 10.01 mmol-H