Merck
  • In vitro toxicological assessment of an organosulfur compound from Allium extract: Cytotoxicity, mutagenicity and genotoxicity studies.
In vitro toxicological assessment of an organosulfur compound from Allium extract: Cytotoxicity, mutagenicity and genotoxicity studies.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2016-12-13)
Pilar Mellado-García, Sara Maisanaba, María Puerto, Ana Isabel Prieto, Ricard Marcos, Silvia Pichardo, Ana María Cameán
ZUSAMMENFASSUNG
Garlic (Allium sativum) and onion (Allium cepa) are being used in the food industry as flavoring but also for their antimicrobial activities. These activities are mainly derived from the organosulfur compounds (OSCs). Propyl propane thiosulfinate (PTS) is an OSC with potential use in the active packaging, but its safety should be guaranteed before being commercialized. The aim of this work was to investigate for the first time the cytotoxicity of PTS as well as its in vitro mutagenic/genotoxic potential using the following battery of genotoxicity tests:(1)the bacterial reverse-mutation assay in S. typhimurium (Ames test, OECD 471, 1997); (2) the micronucleus test (MN, OECD 487, 2016); (3) the mouse lymphoma thymidine-kinase assay (MLA, OECD 476, 2015), and (4) the comet assay (standard and modified with restriction enzymes). The results revealed that PTS was not mutagenic neither in the Ames test nor in MLA. However, genotoxic effects were recorded in the MN test on mammalian cells (L5178YTk
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Aminofluoren, 98%