Merck
Redox Route from Inorganic Precursor Li
ACS nano (2017-02-12)
Paul Simon, Xian-Juan Feng, Matej Bobnar, Peter Höhn, Ulrich Schwarz, Wilder Carrillo-Cabrera, Michael Baitinger, Yuri Grin
ZUSAMMENFASSUNG
We present the synthesis route to carbon with hierarchical morphology on the nanoscale. The structures are generated using crystalline orthorhombic lithium carbide (Li
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Allylmethylsulfon, 96%