Merck
  • Polyvinyl alcohol composite nanofibres containing conjugated levofloxacin-chitosan for controlled drug release.
Polyvinyl alcohol composite nanofibres containing conjugated levofloxacin-chitosan for controlled drug release.
Materials science & engineering. C, Materials for biological applications (2017-02-12)
Javid Jalvandi, Max White, Yuan Gao, Yen Bach Truong, Rajiv Padhye, Ilias Louis Kyratzis
ZUSAMMENFASSUNG
A range of biodegradable drug-nanofibres composite mats have been reported as drug delivery systems. However, their main disadvantage is the rapid release of the drug immediately after application. This paper reports an improved system based on the incorporation of drug conjugated-chitosan into polyvinyl alcohol (PVA) nanofibers. The results showed that controlled release of levofloxacin (LVF) could be achieved by covalently binding LVF to low molecular weight chitosan (CS) via a cleavable amide bond and then blending the conjugated CS with polyvinyl alcohol (PVA) nanofibres prior to electrospinning. PVA/LVF and PVA-CS/LVF nanofibres were fabricated as controls. The conjugated CS-LVF was characterized by FTIR, DSC, TGA and
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethyl-carbodiimid, ≥97.0% (T)