- All-in-one dual-aptasensor capable of rapidly quantifying carcinoembryonic antigen.
All-in-one dual-aptasensor capable of rapidly quantifying carcinoembryonic antigen.
Biosensors & bioelectronics (2016-11-23)
Harriet Khang, Kelly Cho, Stephanie Chong, Ji Hoon Lee
PMID27875751
ZUSAMMENFASSUNG
Using a dual DNA aptamer (CEA aptamer linked to hemin aptamer), capable of rapidly capturing carcinoembryonic antigen (CEA) and hemin, an all-in-one dual-aptasensor with 1,1'-oxalyldiimidazole (ODI) chemiluminescence detection was developed for the early diagnosis of human cancer. CEA and hemin competitively bound with the dual DNA aptamer while the mixture in a detection cell was incubated for 30min at room temperature. When Amplex Red and H