Merck
Small RNA-directed epigenetic programming of embryonic stem cell cardiac differentiation.
Scientific reports (2017-02-07)
Hossein Ghanbarian, Nicole Wagner, Jean-François Michiels, François Cuzin, Kay-Dietrich Wagner, Minoo Rassoulzadegan
ZUSAMMENFASSUNG
Microinjection of small noncoding RNAs in one-cell embryos was reported in several instances to result in transcriptional activation of target genes. To determine the molecular mechanisms involved and to explore whether such epigenetic regulations could play a role in early development, we used a cell culture system as close as possible to the embryonic state. We report efficient cardiac differentiation of embryonic stem (ES) cells induced by small non-coding RNAs with sequences of Cdk9, a key player in cardiomyocyte differentiation. Transfer of oligoribonucleotides representing parts of the Cdk9 mRNA into ES and mouse embryo fibroblast cultures resulted in upregulation of transcription. Dependency on Argonaute proteins and endogenous antisense transcripts indicated that the inducer oligoribonucleotides were processed by the RNAi machinery. Upregulation of Cdk9 expression resulted in increased efficiency of cardiac differentiation suggesting a potential tool for stem cell-based regenerative medicine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus gegen, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Chromatin-Immunpräzipitation (ChIP)-Assay-Kit, Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads.