Merck
  • Physiological Responses of Salinity-Stressed Vibrio sp. and the Effect on the Biofilm Formation on a Nanofiltration Membrane.
Physiological Responses of Salinity-Stressed Vibrio sp. and the Effect on the Biofilm Formation on a Nanofiltration Membrane.
Environmental science & technology (2016-12-21)
Lan Hee Kim, Tzyy Haur Chong
ZUSAMMENFASSUNG
This study evaluated the effects of salinity on the physiological characteristics of Vibrio sp. B2 and biofilm formation on nanofiltration (NF) membrane coupons used in the high recovery seawater desalination process. The test conditions were at 0.6, 1.2, and 2.4 M sodium chloride (NaCl), equivalent to salinity of seawater, brine at 50% and 75% water recovery, respectively. High salinity inhibited the cell growth rate but increased the viability and bacterial membrane integrity. In addition, protein and eDNA concentrations of salinity-stressed bacteria were increased at 1.2 and 2.4 M NaCl. In particular, protein concentration was linearly correlated with the NaCl concentration. Similarly, less biofilm formation on the NF membrane coupon (without permeation flux) was observed by the salinity-stressed bacteria; however, the production of extracellular polymeric substances (EPS) was significantly increased as compared to control, and protein was an influential factor for biofilm formation. This study shows that salinity-stressed bacteria have a high potential to cause biofouling on membrane surface as the bacteria still maintain the cell activity and overproduce EPS. The potential of biofilm formation by the salinity-stressed bacteria has not been reported. Therefore, the findings are important to understand the mechanisms of membrane biofouling in a high salinity environment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)