- Anti-inflammatory activity of Punica granatum L. (Pomegranate) rind extracts applied topically to ex vivo skin.
Anti-inflammatory activity of Punica granatum L. (Pomegranate) rind extracts applied topically to ex vivo skin.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2016-11-22)
David M J Houston, Joachim Bugert, Stephen P Denyer, Charles M Heard
PMID27867111
ZUSAMMENFASSUNG
Coadministered pomegranate rind extract (PRE) and zinc (II) produces a potent virucidal activity against Herpes simplex virus (HSV); however, HSV infections are also associated with localised inflammation and pain. Here, the objective was to determine the anti-inflammatory activity and relative depth penetration of PRE, total pomegranate tannins (TPT) and zinc (II) in skin, ex vivo. PRE, TPT and ZnSO
MATERIALIEN