Merck
  • Developmental Expression Patterns of KCC2 and Functionally Associated Molecules in the Human Brain.
Developmental Expression Patterns of KCC2 and Functionally Associated Molecules in the Human Brain.
Cerebral cortex (New York, N.Y. : 1991) (2015-10-03)
Goran Sedmak, Nataša Jovanov-Milošević, Martin Puskarjov, Monika Ulamec, Božo Krušlin, Kai Kaila, Miloš Judaš
ZUSAMMENFASSUNG
Work on rodents demonstrated that steep upregulation of KCC2, a neuron-specific Cl
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade