Merck
  • Reveglucosidase alfa (BMN 701), an IGF2-Tagged rhAcid α-Glucosidase, Improves Respiratory Functional Parameters in a Murine Model of Pompe Disease.
Reveglucosidase alfa (BMN 701), an IGF2-Tagged rhAcid α-Glucosidase, Improves Respiratory Functional Parameters in a Murine Model of Pompe Disease.
The Journal of pharmacology and experimental therapeutics (2016-11-20)
Jeffrey Peng, Jill Dalton, Mark Butt, Kristin Tracy, Derek Kennedy, Peter Haroldsen, Rhea Cahayag, Stephen Zoog, Charles A O'Neill, Laurie S Tsuruda
ZUSAMMENFASSUNG
Pompe disease is a rare neuromuscular disorder caused by an acid α-glucosidase (GAA) deficiency resulting in glycogen accumulation in muscle, leading to myopathy and respiratory weakness. Reveglucosidase alfa (BMN 701) is an insulin-like growth factor 2-tagged recombinant human acid GAA (rhGAA) that enhances rhGAA cellular uptake via a glycosylation-independent insulin-like growth factor 2 binding region of the cation-independent mannose-6-phosphate receptor (CI-MPR). The studies presented here evaluated the effects of Reveglucosidase alfa treatment on glycogen clearance in muscle relative to rhGAA, as well as changes in respiratory function and glycogen clearance in respiratory-related tissue in a Pompe mouse model (GAA
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DPH, ≥98% (HPLC)