Merck
Relating rheology and tribology of commercial dairy colloids to sensory perception.
Food & function (2016-10-11)
Laura Laguna, Grace Farrell, Michael Bryant, Ardian Morina, Anwesha Sarkar
ZUSAMMENFASSUNG
This study aims to investigate the relationship between rheological and tribological properties of commercial full fat and fat-free/low fat versions of liquid and soft solid colloidal systems (milk, yoghurt, soft cream cheese) with their sensory properties. Oscillatory measurements (strain, frequency), flow curves and tribological measurements (lubrication behaviour using Stribeck analysis) were conducted. Oral condition was mimicked using artificial saliva at 37
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(dimethylsiloxan), viscosity 1.0 cSt (25 °C)