Merck
  • Deciphering the molecular mechanism responsible for GCaMP6m's Ca2+-dependent change in fluorescence.
Deciphering the molecular mechanism responsible for GCaMP6m's Ca2+-dependent change in fluorescence.
PloS one (2017-02-10)
Lauren M Barnett, Thomas E Hughes, Mikhail Drobizhev
ZUSAMMENFASSUNG
The goal of this work is to determine how GCaMP6m's fluorescence is altered in response to Ca2+-binding. Our detailed spectroscopic study reveals the simplest explanation for how GCaMP6m changes fluorescence in response to Ca2+ is with a four-state model, in which a Ca2+-dependent change of the chromophore protonation state, due to a shift in pKa, is the predominant factor. The pKa shift is quantitatively explained by a change in electrostatic potential around the chromophore due to the conformational changes that occur in the protein when calmodulin binds Ca2+ and interacts with the M13 peptide. The absolute pKa values for the Ca2+-free and Ca2+-saturated states of GCaMP6m are critical to its high signal-to-noise ratio. This mechanism has important implications for further improvements to GCaMP6m and potentially for other similarly designed biosensors.