Merck
  • High efficiency removal of triclosan by structure-directing agent modified mesoporous MIL-53(Al).
High efficiency removal of triclosan by structure-directing agent modified mesoporous MIL-53(Al).
Environmental science and pollution research international (2017-02-19)
Rongni Dou, Junya Zhang, Yuancai Chen, Siyuan Feng
ZUSAMMENFASSUNG
In order to expand the potential applications of metal-organic frameworks (MOFs), structure directing agents modified mesoporous MIL-53(Al) (MIL-53(Al)-1) was investigated to adsorb triclosan (TCS) with two different initial concentrations. MIL-53(Al)-1 with high mesoporosity and total pore volume exhibited higher adsorption capacity and 4.4 times faster adsorption of TCS at low concentration (1 mg L
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,1,1-Trifluor-5,5-dimethyl-2,4-hexandion, 98%