Merck
  • Restoring the impaired cardiac calcium homeostasis and cardiac function in iron overload rats by the combined deferiprone and N-acetyl cysteine.
Restoring the impaired cardiac calcium homeostasis and cardiac function in iron overload rats by the combined deferiprone and N-acetyl cysteine.
Scientific reports (2017-03-14)
Suwakon Wongjaikam, Sirinart Kumfu, Juthamas Khamseekaew, Siriporn C Chattipakorn, Nipon Chattipakorn
ZUSAMMENFASSUNG
Intracellular calcium [Ca
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ferrocen, 98%
Sigma-Aldrich
Anti-Calcium Channel (α1C Subunit) (L-type of Voltage-gated Ca2+ Channel) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)