Merck
  • Ultrasensitive electrochemical aptasensor based on sandwich architecture for selective label-free detection of colorectal cancer (CT26) cells.
Ultrasensitive electrochemical aptasensor based on sandwich architecture for selective label-free detection of colorectal cancer (CT26) cells.
Biosensors & bioelectronics (2016-11-11)
Ayemeh Bagheri Hashkavayi, Jahan Bakhsh Raoof, Reza Ojani, Saeid Kavoosian
ZUSAMMENFASSUNG
Colorectal cancer is one of the most common cancers in the world and has no effective treatment. Therefore, development of new methods for early diagnosis is instantly required. Biological recognition probes such as synthetic receptor and aptamer is one of the candidate recognition layers to detect important biomolecules. In this work, an electrochemical aptasensor was developed by fabricating an aptamer-cell-aptamer sandwich architecture on an SBA-15-3-aminopropyltriethoxysilane (SBA-15-pr-NH
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ruthenium(III)-chlorid, Anhydrous, solid, Ru content 45-55%