Merck
  • Glycated lysine-141 in haptoglobin improves the diagnostic accuracy for type 2 diabetes mellitus in combination with glycated hemoglobin HbA
Glycated lysine-141 in haptoglobin improves the diagnostic accuracy for type 2 diabetes mellitus in combination with glycated hemoglobin HbA
Clinical proteomics (2017-04-01)
Sandro Spiller, Yichao Li, Matthias Blüher, Lonnie Welch, Ralf Hoffmann
ZUSAMMENFASSUNG
Recent epidemiological studies indicate that only 30-50% of undiagnosed type 2 diabetes mellitus (T2DM) patients are identified using glycated hemoglobin (HbA Here, we studied the glycation degrees of 27 glycation sites representing nine plasma proteins in 48 newly diagnosed male T2DM patients and 48 non-diabetic men matched for age (range 35-65 years). Samples were digested with trypsin and enriched for glycated peptides using boronic acid affinity chromatography. Quantification relied on mass spectrometry (multiple reaction monitoring) using isotope-labelled peptides as internal standard. The combination of glycated lysine-141 of haptoglobin (HP K141) and HbA Our studies demonstrated the great potential of glycation sites in plasma proteins providing an additional diagnostic tool for T2DM and elucidating that the combination of these sites with HbA
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Methioninsulfoxid
Sigma-Aldrich
DL-Cystein, technical grade