Merck
  • Physicochemical Property and Oxidative Stability of Whey Protein Concentrate Multiple Nanoemulsion Containing Fish Oil.
Physicochemical Property and Oxidative Stability of Whey Protein Concentrate Multiple Nanoemulsion Containing Fish Oil.
Journal of food science (2017-01-20)
Jae-Young Hwang, Ho-Kyung Ha, Mee-Ryung Lee, Jin Wook Kim, Hyun-Jin Kim, Won-Jae Lee
ZUSAMMENFASSUNG
The objectives of this research were to produce whey protein concentrate (WPC) multiple nanoemulsion (MNE) and to study how whey protein concentration level and antioxidant type affected the physicochemical properties and oxidative stability of fish oil in MNE. The morphological and physicochemical characteristics of MNE were investigated by using transmission electron microscopy and particle size analyzer, respectively. The oxidative stability of fish oil in MNEs was assessed by measuring peroxide value (PV), p-anisidine value, and volatile compounds. The spherical forms of emulsions with size ranging from 190 to 210 nm were observed indicating the successful production of MNE. Compared with free fish oil, fish oil in MNE exhibited lower PV, p-anisidine value, and formation of maker of oxidation of fish oil indicating the oxidative stability of fish oil in MNE was enhanced. PV, p-anisidine value, and makers of oxidation of fish oil were decreased with increased WPC concentration level. The combined use of Vitamin C and E in MNE resulted in a reduction in PV and p-anisidine value, and development of maker of oxidation. In conclusion, WPC concentration level and antioxidant type are key factors affecting the droplet size of MNE and oxidative stability of fish oil.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumthiocyanat, ACS reagent, ≥97.5%
Sigma-Aldrich
p-Anisidin, 99%
Sigma-Aldrich
Cumolhydroperoxid, technical grade, 80%