Merck
Intestinal epithelial organoids fuse to form self-organizing tubes in floating collagen gels.
Development (Cambridge, England) (2017-03-16)
Norman Sachs, Yoshiyuki Tsukamoto, Pekka Kujala, Peter J Peters, Hans Clevers
ZUSAMMENFASSUNG
Multiple recent examples highlight how stem cells can self-organize
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade