Merck
  • Usnic Acid Activity on Oxidative and Nitrosative Stress of Azole-Resistant Candida albicans Biofilm.
Usnic Acid Activity on Oxidative and Nitrosative Stress of Azole-Resistant Candida albicans Biofilm.
Planta medica (2016-09-21)
Mariana Andrea Peralta, María Angel da Silva, María Gabriela Ortega, José Luis Cabrera, María Gabriela Paraje
ZUSAMMENFASSUNG
Several studies report that (+)-usnic acid, a lichen secondary metabolite, inhibits growth of different bacteria and fungi; however, the mechanism of its antimicrobial activity remains unknown. In this study, we explored the ability of usnic acid, obtained from
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluconazol, ≥98% (HPLC), powder