Merck
  • The sphingosine 1-phosphate receptor 1 causes tissue retention by inhibiting the entry of peripheral tissue T lymphocytes into afferent lymphatics.
The sphingosine 1-phosphate receptor 1 causes tissue retention by inhibiting the entry of peripheral tissue T lymphocytes into afferent lymphatics.
Nature immunology (2007-11-27)
Levi G Ledgerwood, Girdhari Lal, Nan Zhang, Alexandre Garin, Steven J Esses, Florent Ginhoux, Miriam Merad, Helene Peche, Sergio A Lira, Yaozhong Ding, Yu Yang, Xingxuan He, Edward H Schuchman, Maria L Allende, Jordi C Ochando, Jonathan S Bromberg
ZUSAMMENFASSUNG
Although much is known about the migration of T cells from blood to lymph nodes, less is known about the mechanisms regulating the migration of T cells from tissues into lymph nodes through afferent lymphatics. Here we investigated T cell egress from nonlymphoid tissues into afferent lymph in vivo and developed an experimental model to recapitulate this process in vitro. Agonism of sphingosine 1-phosphate receptor 1 inhibited the entry of tissue T cells into afferent lymphatics in homeostatic and inflammatory conditions and caused the arrest, mediated at least partially by interactions of the integrin LFA-1 with its ligand ICAM-1 and of the integrin VLA-4 with its ligand VCAM-1, of polarized T cells at the basal surface of lymphatic but not blood vessel endothelium. Thus, the increased sphingosine 1-phosphate present in inflamed peripheral tissues may induce T cell retention and suppress T cell egress.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
PKH67 grün fluoreszierendes Zelllinker-Minikit für die allgemeine Zellmembranmarkierung, Distributed for Phanos Technologies
Sigma-Aldrich
PKH67 grün fluoreszierendes Zelllinker-Kit für die allgemeine Zellmembranmarkierung, Distributed for Phanos Technologies
Sigma-Aldrich
PKH67 grün fluoreszierendes Zelllinker-Midikit für die allgemeine Zellmembranmarkierung, Distributed for Phanos Technologies