- Cyclin F/FBXO1 Interacts with HIV-1 Viral Infectivity Factor (Vif) and Restricts Progeny Virion Infectivity by Ubiquitination and Proteasomal Degradation of Vif Protein through SCF
Cyclin F/FBXO1 Interacts with HIV-1 Viral Infectivity Factor (Vif) and Restricts Progeny Virion Infectivity by Ubiquitination and Proteasomal Degradation of Vif Protein through SCF
The Journal of biological chemistry (2017-02-12)
Tracy Augustine, Priyanka Chaudhary, Kailash Gupta, Sehbanul Islam, Payel Ghosh, Manas Kumar Santra, Debashis Mitra
PMID28184007
ZUSAMMENFASSUNG
Cyclin F protein, also known as FBXO1, is the largest among all cyclins and oscillates in the cell cycle like other cyclins. Apart from being a G
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)