- Photosensitizer-eluting nanofibers for enhanced photodynamic therapy of wounds: A preclinical study in immunocompromized rats.
Photosensitizer-eluting nanofibers for enhanced photodynamic therapy of wounds: A preclinical study in immunocompromized rats.
International journal of pharmaceutics (2017-02-07)
Labiba El-Khordagui, Nesma El-Sayed, Sally Galal, Hanan El-Gowelli, Hoda Omar, Moustafa Mohamed
PMID28163229
ZUSAMMENFASSUNG
Electrospun nanofibers (NFs) as drug delivery/tissue regeneration template and antimicrobial photodynamic therapy (APDT) have been widely investigated as two different approaches to enhance wound healing. In the present study, the two approaches were combined in a single platform for greater healing enhancement potentials. Composite photosensitizer-eluting NFs were developed using a polyhydrohybutyrate/polyethylene glycol (60:40 PHB/PEG) polymer blend and methylene blue (MB) as antimicrobial photosensitizer (PS). NFs protected the photoactivity of entrapped MB, enhanced its photodynamic activity against two wound bacteria, Staphylococcus aureus standard strain (SA
MATERIALIEN