Merck
LPA
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-01-11)
Péter Tibor Dancs, Éva Ruisanchez, Andrea Balogh, Cecília Rita Panta, Zsuzsanna Miklós, Rolf M Nüsing, Junken Aoki, Jerold Chun, Stefan Offermanns, Gábor Tigyi, Zoltán Benyó
ZUSAMMENFASSUNG
Lysophosphatidic acid (LPA) has been recognized recently as an endothelium-dependent vasodilator, but several lines of evidence indicate that it may also stimulate vascular smooth muscle cells (VSMCs), thereby contributing to vasoregulation and remodeling. In the present study, mRNA expression of all 6 LPA receptor genes was detected in murine aortic VSMCs, with the highest levels of LPA