Merck
  • Human and soil exposure during mechanical chlorpyrifos, myclobutanil and copper oxychloride application in a peach orchard in Argentina.
Human and soil exposure during mechanical chlorpyrifos, myclobutanil and copper oxychloride application in a peach orchard in Argentina.
The Science of the total environment (2017-02-27)
Giselle Berenstein, Soledad Nasello, Érica Beiguel, Pedro Flores, Johanna Di Schiena, Silvana Basack, Enrique A Hughes, Anita Zalts, Javier M Montserrat
ZUSAMMENFASSUNG
The objective of this study was to measure the impact of the mechanized chlorpyrifos, copper oxychloride and myclobutanil application in a small peach orchard, on humans (operators, bystanders and residents) and on the productive soil. The mean Potential Dermal Exposure (PDE) of the workers (tractor drivers) was 30.8mL·h
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Myclobutanil, PESTANAL®, analytical standard
Sigma-Aldrich
1,1,1-Trifluor-5,5-dimethyl-2,4-hexandion, 98%