- Genetic and Dietary Iron Overload Differentially Affect the Course of
Genetic and Dietary Iron Overload Differentially Affect the Course of
Frontiers in cellular and infection microbiology (2017-04-27)
Manfred Nairz, Andrea Schroll, David Haschka, Stefanie Dichtl, Piotr Tymoszuk, Egon Demetz, Patrizia Moser, Hubertus Haas, Ferric C Fang, Igor Theurl, Günter Weiss
PMID28443246
ZUSAMMENFASSUNG
Genetic and dietary forms of iron overload have distinctive clinical and pathophysiological features. HFE-associated hereditary hemochromatosis is characterized by overwhelming intestinal iron absorption, parenchymal iron deposition, and macrophage iron depletion. In contrast, excessive dietary iron intake results in iron deposition in macrophages. However, the functional consequences of genetic and dietary iron overload for the control of microbes are incompletely understood. Using