Merck
  • Surfactant protein A (SP-A) and SP-A-derived peptide attenuate chemotaxis of mast cells induced by human β-defensin 3.
Surfactant protein A (SP-A) and SP-A-derived peptide attenuate chemotaxis of mast cells induced by human β-defensin 3.
Biochemical and biophysical research communications (2017-02-12)
Yasuaki Uehara, Motoko Takahashi, Masaki Murata, Atsushi Saito, Rina Takamiya, Yoshihiro Hasegawa, Koji Kuronuma, Hirofumi Chiba, Jiro Hashimoto, Norimasa Sawada, Hiroki Takahashi, Yoshio Kuroki, Shigeru Ariki
ZUSAMMENFASSUNG
Human β-defensin 3 (hBD3) is known to be involved in mast cell activation. However, molecular mechanisms underlying the regulation of hBD3-induced mast cell activation have been poorly understood. We previously reported that SP-A and SP-A-derived peptide 01 (SAP01) regulate the function of hBD3. In this study, we focused on the effects of SP-A and SAP01 on the activation of mast cells induced by hBD3. SAP01 directly bound to hBD3. Mast cell-mediated vascular permeability and edema in hBD3 administered rat ears were decreased when injected with SP-A or SAP01. Compatible with the results in rat ear model, both SP-A and SAP01 inhibited hBD3-induced chemotaxis of mast cells in vitro. Direct interaction between SP-A or SAP01 and hBD3 seemed to be responsible for the inhibitory effects on chemotaxis. Furthermore, SAP01 attenuated hBD3-induced accumulation of mast cells and eosinophils in tracheas of the OVA-sensitized inflammatory model. SP-A might contribute to the regulation of inflammatory responses mediated by mast cells during infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Toluidinblau, suitable for microscopy, 8.74% (ZN (THEORY))