- Human Keratinocytes Respond to Extracellular UTP by Induction of Hyaluronan Synthase 2 Expression and Increased Hyaluronan Synthesis.
Human Keratinocytes Respond to Extracellular UTP by Induction of Hyaluronan Synthase 2 Expression and Increased Hyaluronan Synthesis.
The Journal of biological chemistry (2017-02-12)
Tiina Jokela, Riikka Kärnä, Leena Rauhala, Genevieve Bart, Sanna Pasonen-Seppänen, Sanna Oikari, Markku I Tammi, Raija H Tammi
PMID28188289
ZUSAMMENFASSUNG
The release of nucleotides into extracellular space is triggered by insults like wounding and ultraviolet radiation, resulting in stimulatory or inhibitory signals via plasma membrane nucleotide receptors. As similar insults are known to activate hyaluronan synthesis we explored the possibility that extracellular UTP or its breakdown products UDP and UMP act as mediators for hyaluronan synthase (
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, niedriger Glucosegehalt, With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.
Sigma-Aldrich
Bisindolylmaleimid I, A highly selective, cell-permeable, and reversible protein kinase C (PKC) inhibitor (IC50 = 10 nM) that is structurally similar to staurosporine.
Sigma-Aldrich
KN-93, A cell-permeable, reversible and competitive inhibitor of rat brain CaM kinase II (Ki = 370 nM).