- Removal of gadolinium-based contrast agents: adsorption on activated carbon.
Removal of gadolinium-based contrast agents: adsorption on activated carbon.
Environmental science and pollution research international (2017-02-02)
María P Elizalde-González, Esmeralda García-Díaz, Mario González-Perea, Jürgen Mattusch
PMID28144867
ZUSAMMENFASSUNG
Three carbon samples were employed in this work, including commercial (1690 m