Merck
Non-Adrenergic, Tamsulosin-Insensitive Smooth Muscle Contraction is Sufficient to Replace α
The Prostate (2017-01-25)
Martin Hennenberg, Alice Acevedo, Nicolas Wiemer, Aysenur Kan, Alexander Tamalunas, Yiming Wang, Qingfeng Yu, Beata Rutz, Anna Ciotkowska, Annika Herlemann, Frank Strittmatter, Christian G Stief, Christian Gratzke
ZUSAMMENFASSUNG
Lower urinary tract symptoms (LUTS) suggestive of benign prostatic hyperplasia may be caused by prostate smooth muscle contraction. Although α Prostate tissues were obtained from radical prostatectomy (n = 127 patients). Contractile responses were studied in an organ bath. Endothelin-1 and noradrenaline induced contractions of similar magnitude (116 ± 23 and 117 ± 18% of KCl-induced contractions). Endothelin-2- and -3-induced maximum contractions of 63 ± 8.6 and 71 ± 19% of KCl, while contractions by the thromboxane analog U46619 amounted up to 63 ± 9.4%. Dopamine-induced contractions averaged to 22 ± 4.5% of KCl, while maximum contractions by serotonin, histamine, and carbachol stayed below 10% of KCl-induced. While noradrenaline-induced contractions were inhibited by tamsulosin (300 nM), endothelin-1-, -2-, or -3-induced contraction were not. No additive effects were observed if endothelins and noradrenaline were applied consecutively to the same samples. If endothelin-1 was applied after U46619, resulting tension (172 ± 43% of KCl) significantly exceeded noradrenaline-induced contraction. Tensions following combined application of endothelin-2 or -3 with U46619 stayed below noradrenaline-induced contractions. Tension following combined application of all three endothelins with U46619 resembled maximum noradrenaline-induced tone. Contractions following concomitant confrontation of human prostate tissue with noradrenaline and endothelin-1 are not additive. Endothelin-1 is sufficient to induce a smooth muscle tone resembling that of noradrenaline. This may replace lacking α
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Serotonin -hydrochlorid, powder
Sigma-Aldrich
L-Norepinephrin -hydrochlorid, ≥98.0% (sum of enantiomers, HPLC)