- An experimental examination of interindividual variation in feather corticosterone content in the house sparrow, Passer domesticus in southeast Australia.
An experimental examination of interindividual variation in feather corticosterone content in the house sparrow, Passer domesticus in southeast Australia.
General and comparative endocrinology (2015-12-25)
Yaara Aharon-Rotman, Katherine L Buchanan, Marcel Klaassen, William A Buttemer
PMID26699204
ZUSAMMENFASSUNG
Non-invasive techniques for measuring glucocorticoids (GCs) have become more prevalent, due to the advantage of eliminating the effects of animal disturbance on GC levels and their potential to provide an integrated, historic estimate of circulating GC levels. In the case of birds, corticosterone (CORT) is deposited in feathers, and may reflect a bird's GC status over the period of feather synthesis. This technique thus permits a retrospective view of the average circulating GC levels during the moult period. While it is generally assumed that differences in feather CORT content (CORT