Merck
Endogenous GAB
Oncogene (2016-07-05)
A Blanchart, R Fernando, M Häring, N Assaife-Lopes, R A Romanov, M Andäng, T Harkany, P Ernfors
ZUSAMMENFASSUNG
Although genome alterations driving glioma by fueling cell malignancy have largely been resolved, less is known of the impact of tumor environment on disease progression. Here, we demonstrate functional GABA