Merck
  • Tetraspanin 6: a pivotal protein of the multiple vesicular body determining exosome release and lysosomal degradation of amyloid precursor protein fragments.
Tetraspanin 6: a pivotal protein of the multiple vesicular body determining exosome release and lysosomal degradation of amyloid precursor protein fragments.
Molecular neurodegeneration (2017-03-11)
Francesc X Guix, Ragna Sannerud, Fedor Berditchevski, Amaia M Arranz, Katrien Horré, An Snellinx, Amantha Thathiah, Takaomi Saido, Takashi Saito, Sundaresan Rajesh, Michael Overduin, Samir Kumar-Singh, Enrico Radaelli, Nikky Corthout, Julien Colombelli, Sébastien Tosi, Sebastian Munck, Isabel H Salas, Wim Annaert, Bart De Strooper
ZUSAMMENFASSUNG
The mechanisms behind Aβ-peptide accumulation in non-familial Alzheimer's disease (AD) remain elusive. Proteins of the tetraspanin family modulate Aβ production by interacting to γ-secretase. We searched for tetraspanins with altered expression in AD brains. The function of the selected tetraspanin was studied in vitro and the physiological relevance of our findings was confirmed in vivo. Tetraspanin-6 (TSPAN6) is increased in AD brains and overexpression in cells exerts paradoxical effects on Amyloid Precursor Protein (APP) metabolism, increasing APP-C-terminal fragments (APP-CTF) and Aβ levels at the same time. TSPAN6 affects autophagosome-lysosomal fusion slowing down the degradation of APP-CTF. TSPAN6 recruits also the cytosolic, exosome-forming adaptor syntenin which increases secretion of exosomes that contain APP-CTF. TSPAN6 is a key player in the bifurcation between lysosomal-dependent degradation and exosome mediated secretion of APP-CTF. This corroborates the central role of the autophagosomal/lysosomal pathway in APP metabolism and shows that TSPAN6 is a crucial player in APP-CTF turnover.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Presenilin-1-Antikörper, NT, Klon hPS1-NT, culture supernatant, clone hPS1-NT, Chemicon®
Sigma-Aldrich
MISSION® esiRNA, targeting human TSPAN6