Merck
Functional and Structural Characterization of a Thermostable Phospholipase A
Journal of agricultural and food chemistry (2017-03-14)
Nabil Smichi, Houcemeddine Othman, Neila Achouri, Alexandre Noiriel, Vincent Arondel, Najet Srairi-Abid, Abdelkarim Abousalham, Youssef Gargouri, Nabil Miled, Ahmed Fendri
ZUSAMMENFASSUNG
Novel phospholipase (PLA
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,2-Didodecanoyl-sn-Glycero-3-Phosphocholin, ≥99%, synthetic