- Prediction and design of efficient exciplex emitters for high-efficiency, thermally activated delayed-fluorescence organic light-emitting diodes.
Prediction and design of efficient exciplex emitters for high-efficiency, thermally activated delayed-fluorescence organic light-emitting diodes.
Advanced materials (Deerfield Beach, Fla.) (2015-02-26)
Xiao-Ke Liu, Zhan Chen, Cai-Jun Zheng, Chuan-Lin Liu, Chun-Sing Lee, Fan Li, Xue-Mei Ou, Xiao-Hong Zhang
PMID25712786
ZUSAMMENFASSUNG
High-efficiency, thermally activated delayed-fluorescence organic light-emitting diodes based on exciplex emitters are demonstrated. The best device, based on a TAPC:DPTPCz emitter, shows a high external quantum efficiency of 15.4%. Strategies for predicting and designing efficient exciplex emitters are also provided. This approach allow prediction and design of efficient exciplex emitters for achieving high-efficiency organic light-emitting diodes, for future use in displays and lighting applications.