Merck
RESOLUTION OF NONINFECTIOUS UVEITIC CYSTOID MACULAR EDEMA WITH TOPICAL DIFLUPREDNATE.
Retina (Philadelphia, Pa.) (2016-08-17)
Daniel L Feiler, Sunil K Srivastava, Francesco Pichi, James Bena, Careen Y Lowder
ZUSAMMENFASSUNG
To evaluate the short-term safety and efficacy of topical difluprednate (0.05%) for the treatment of noninfectious uveitic cystoid macular edema. Twenty-seven patients (35 eyes) undergoing treatment with difluprednate 4 times daily for 3 weeks for noninfectious uveitic cystoid macular edema were reviewed for visual acuity, intraocular pressure, optical coherence tomography, and fluorescein angiography results. A mixed model analysis was fit with each measure as the outcome, visit as the primary predictor, and patient and eye as random effects. Mean central foveal thickness decreased by 117 μm (P < 0.001) at 30 ± 15 days, 124 μm (P < 0.001) at 60 ± 15 days, and 152 μm (P < 0.001) at 180 ± 30 days. Complete resolution of intraretinal fluid was observed in 15 of 34 (44%) eyes at 30 ± 15 days, 11 of 21 (52%) eyes at 60 ± 15 days, and 9 of 12 (75%) eyes at 180 ± 30 days. Improvement in fluorescein leakage was noted in 7 of 8 eyes (88%). Visual acuity improved by a mean of 5 letters (P = 0.001) at 30 ± 15 days, 5.5 letters (P = 0.007) at 60 ± 15 days, and 7 letters (P = 0.032) at 180 ± 30 days. Mean increase in intraocular pressure was 1.48 mmHg at 30 ± 15 days (P = 0.080), 1.92 mmHg at 60 ± 15 days (P = 0.110), and 6.18 mmHg (P = 0.001) at 180 ± 30 days. Topical difluprednate is a well-tolerated and effective treatment for noninfectious uveitic cystoid macular edema with decreased central foveal thickness, mild improvement in visual acuity, and elevation of intraocular pressure observed in a few patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6α,9α-Difluoroprednisolone 21-acetate 17-butyrate, ≥98%