- Development and validation of a liquid chromatography tandem mass spectrometry assay for the measurement of faecal metronidazole.
Development and validation of a liquid chromatography tandem mass spectrometry assay for the measurement of faecal metronidazole.
Clinical biochemistry (2016-12-10)
Jinny Jeffery, Zoe J Vincent, Ruth M Ayling, Stephen J Lewis
PMID27932283
ZUSAMMENFASSUNG
Metronidazole is an oral antibiotic which is widely used in the treatment of patients with Clostridium difficile associated disease. This article describes the validation of a LC-MS/MS assay for the measurement of metronidazole in human faecal samples. Matrix matched and aqueous standards showed no significant difference in performance for the routine calibration of the assay. D Metronidazole can be quantified in faecal samples using LC-MS/MS which opens up opportunities for further research in this area.