Merck
Lipoic and dihydrolipoic acids as antioxidants. A critical evaluation.
Free radical research (1994-02-01)
B C Scott, O I Aruoma, P J Evans, C O'Neill, A Van der Vliet, C E Cross, H Tritschler, B Halliwell
ZUSAMMENFASSUNG
A detailed evaluation of the antioxidant and pro-oxidant properties of lipoic acid (LA) and dihydrolipoic acid (DHLA) was performed. Both compounds are powerful scavengers of hypochlorous acid, able to protect alpha 1-antiproteinase against inactivation by HOCl. LA was a powerful scavenger of hydroxyl radicals (OH.) and could inhibit both iron-dependent OH. generation and peroxidation of ox-brain phospholipid liposomes in the presence of FeCl3-ascorbate, presumably by binding iron ions and rendering them redox-inactive. By contrast, DHLA accelerated iron-dependent OH. generation and lipid peroxidation, probably by reducing Fe3+ to Fe2+. LA inhibited this pro-oxidant action of DHLA. However, DHLA did not accelerate DNA degradation by a ferric bleomycin complex and slightly inhibited peroxidation of arachidonic acid by the myoglobin-H2O2 system. Under certain circumstances, DHLA accelerated the loss of activity of alpha-antiproteinase exposed to ionizing radiation under a N2O/O2 atmosphere and also the loss of creatine kinase activity in human plasma exposed to gas-phase cigarette smoke. Neither LA nor DHLA reacted with superoxide radical (O.2-) or H2O2 at significant rates, but both were good scavengers of trichloromethylperoxyl radical (CCl3O2.). We conclude that LA and DHLA have powerful antioxidant properties. However, DHLA can also exert pro-oxidant properties, both by its iron ion-reducing ability and probably by its ability to generate reactive sulphur-containing radicals that can damage certain proteins, such as alpha 1-antiproteinase and creatine kinase.