Merck
AIE active turn-off fluorescent probe for the detection of Cu
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-04-25)
Mehboobali Pannipara, Abdullah G Al-Sehemi, Abul Kalam, Abdullah M Asiri, Muhammad Nadeem Arshad
ZUSAMMENFASSUNG
A novel AIE active Schiff base compound (receptor 1) has been designed, synthesized and characterized spectroscopically. Receptor 1 show weak emission in solution state but emit strongly in solid state. The investigation on the AIE behavior of receptor 1 using solvent-non-solvent (THF-Water) interaction reveals a maximum PL intensity on reaching the water fraction 90% (fw=90) due to nanoaggregation. Furthermore, demonstration of metal ion sensing application of receptor 1 unravel that it could act as selective and sensitive sensor for Cu
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Hydroxy-1-Naphthaldehyd, technical grade