Merck
  • Anionically functionalized guar gum embedded with silica nanoparticles: An efficient nanocomposite adsorbent for rapid adsorptive removal of toxic cationic dyes and metal ions.
Anionically functionalized guar gum embedded with silica nanoparticles: An efficient nanocomposite adsorbent for rapid adsorptive removal of toxic cationic dyes and metal ions.
Bioresource technology (2016-12-14)
Abhay Shankar Patra, Soumitra Ghorai, Debabrata Sarkar, Raghunath Das, Supriya Sarkar, Sagar Pal
ZUSAMMENFASSUNG
In the present work, a novel biodegradable nanocomposite has been developed (h-GG/SiO