Merck
  • COMT Val158Met Polymorphism and Symptom Improvement Following a Cognitively Focused Intervention for Irritable Bowel Syndrome.
COMT Val158Met Polymorphism and Symptom Improvement Following a Cognitively Focused Intervention for Irritable Bowel Syndrome.
Nursing research (2017-03-03)
Claire J Han, Ruth Kohen, Sangeun Jun, Monica E Jarrett, Kevin C Cain, Robert Burr, Margaret M Heitkemper
ZUSAMMENFASSUNG
Our nurse-delivered comprehensive self-management (CSM) program, a cognitive behavioral therapy intervention, is effective in reducing gastrointestinal and psychological distress symptoms in patients with irritable bowel syndrome (IBS). Findings from non-IBS studies indicate that the catechol-O-methyltransferase (COMT) Val158Met polymorphism may moderate the efficacy of cognitive behavioral therapy. It is unknown whether this COMT polymorphism is associated with symptom improvements in patients with IBS. We tested whether this COMT Val158Met polymorphism influences the efficacy of our 2-month CSM intervention. We analyzed data from two published randomized controlled trials of CSM. The combined European American sample included 149 women and 23 men with IBS (CSM, n = 111; usual care [UC], n = 61). The primary outcomes were daily reports of abdominal pain, depression, anxiety, and feeling stressed measured 3 and 6 months after randomization. Secondary outcomes were additional daily symptoms, retrospective psychological distress, IBS quality of life, and cognitive beliefs about IBS. The interaction between COMT Val158Met polymorphism and treatment group (CSM vs. UC) in a generalized estimating equation model tested the main objective. At 3 months, participants with at least one Val allele benefited more from CSM than did those with the Met/Met genotype (p = .01 for anxiety and feeling stressed, and p < .16 for abdominal pain and depression). The moderating effect of genotype was weaker at 6 months. Persons with at least one Val allele may benefit more from CSM than those homozygous for the Met allele. Future studies with larger and more racially diverse samples are needed to confirm these findings. Parent studies were registered at ClinicalTrials.gov (NCT00167635 and NCT00907790).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluorescein (freie Säure), Dye content 95 %