Merck
NDV-D90 suppresses growth of gastric cancer and cancer-related vascularization.
Oncotarget (2017-04-08)
Hong Sui, Kaibing Wang, Rui Xie, Xi Li, Kunpeng Li, Yuxian Bai, Xishan Wang, Bin Bai, Dan Chen, Jiazhuang Li, Baozhong Shen
ZUSAMMENFASSUNG
Recent reports suggest promises on using oncolytic Newcastle disease viruses (NDV) to treat different cancers, while the effects of a NDV-D90 strain on gastric cancer remain unknown. Here we showed that NDV-D90 induced gastric cancer cell apoptosis in a dose-dependent manner in 3 gastric cancer cell lines BGC-823, SGC-7901 and MKN-28. Pronounced reduction in cell invasion was detected in NDV-D90-treated BGC-823 and SGC-7901 cells, but not in MKN-28 cells. The increases in cell apoptosis and reduction in cell growth in NDV-D90-treated gastric cancer cells seemingly resulted from augmentation of p38 signaling and suppression of ERK1/2 and Akt signaling. In vivo, orthotopic injection of NDV-D90 impaired tumor growth and induced intratumoral necrosis. Tumor cells that had been pre-treated with NDV-D90 showed defect in development of implanted tumor. Moreover, NDV-D90 appeared to reduce gastric tumor vascularization, possibly through suppression of vascular endothelial growth factor A and Matrix Metallopeptidase 2. Together, our data suggest that NDV-D90 may have potential anti-cancer effects on gastric cancer.