Merck
Structural insights into the ene-reductase synthesis of profens.
Organic & biomolecular chemistry (2017-05-10)
J Waller, H S Toogood, V Karuppiah, N J W Rattray, D J Mansell, D Leys, J M Gardiner, A Fryszkowska, S T Ahmed, R Bandichhor, G P Reddy, N S Scrutton
ZUSAMMENFASSUNG
Reduction of double bonds of α,β-unsaturated carboxylic acids and esters by ene-reductases remains challenging and it typically requires activation by a second electron-withdrawing moiety, such as a halide or second carboxylate group. We showed that profen precursors, 2-arylpropenoic acids and their esters, were efficiently reduced by Old Yellow Enzymes (OYEs). The XenA and GYE enzymes showed activity towards acids, while a wider range of enzymes were active towards the equivalent methyl esters. Comparative co-crystal structural analysis of profen-bound OYEs highlighted key interactions important in determining substrate binding in a catalytically active conformation. The general utility of ene reductases for the synthesis of (R)-profens was established and this work will now drive future mutagenesis studies to screen for the production of pharmaceutically-active (S)-profens.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Phenylpropionsäure, 97%