Merck
  • Sulfatase-2 promotes the growth and metastasis of colorectal cancer by activating Akt and Erk1/2 pathways.
Sulfatase-2 promotes the growth and metastasis of colorectal cancer by activating Akt and Erk1/2 pathways.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2017-03-23)
Youmao Tao, Tao Han, Tao Zhang, Caixia Sun
ZUSAMMENFASSUNG
The molecular mechanisms underlying the growth and metastasis of colorectal cancer (CRC) remain largely unknown. Sulfatase-2 (SULF2) was found to play critical roles in human cancers. Recent study reported that SULF1/2 overexpression resulted in increased viability and proliferation, and augmented cell migration in CRC cells. However, the expression of SULF2 and its underlying molecular mechanisms in CRC remain unknown. In this study, we found that the expressions of SULF2 in CRC tissues and cell lines were significantly increased compared to control groups. Increased expression of SULF2 was associated with malignant clinical features and poor prognosis of CRC patients. Loss of SULF2 significantly prohibited the proliferation, cell cycle progression, migration and invasion of HT29 cells, while restoration of SULF2 significantly promoted these cellular functions of SW480 cells. In vivo tumorigenicity and liver metastasis assays confirmed that SULF2 knockdown significantly reduced the growth and metastatic abilities of HT29 cells in nude mice. Furthermore, SULF2 knockdown reduced the levels of p-Akt and p-Erk1/2 in HT29 cells, while SULF2 overexpression showed opposite effects on the expressions of these proteins in SW480 cells. In all, SULF2 promotes the growth and metastasis of CRC probably by activating Akt and Erk1/2 pathways. SULF2 potentially serves as a promising biomarker and therapeutic target in CRC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human SULF2