- Strength Training Decreases Inflammation and Increases Cognition and Physical Fitness in Older Women with Cognitive Impairment.
Strength Training Decreases Inflammation and Increases Cognition and Physical Fitness in Older Women with Cognitive Impairment.
Frontiers in physiology (2017-07-01)
Matheus U Chupel, Fábio Direito, Guilherme E Furtado, Luciéle G Minuzzi, Filipa M Pedrosa, Juan C Colado, José P Ferreira, Edith Filaire, Ana M Teixeira
PMID28659812
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bovine IFNG / Interferon Gamma ELISA Kit, for serum, plasma and cell culture supernatants